missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Urtoxazumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541859
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Urtoxazumab is a biosimilar that targets Stx-2.
Specifications
| Urtoxazumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HuVTm1.1; SLT-II; Stx-2; TMA-15 | |
| Escherichia coli Stx-2/SLT-II. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction