missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USAG1/SOSTDC1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98942AF532
This item is not returnable.
View return policy
Description
USAG1/SOSTDC1 Polyclonal antibody specifically detects USAG1/SOSTDC1 in Human samples. It is validated for Western Blot
Specifications
| USAG1/SOSTDC1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| cystine-knot containing secreted protein, DKFZp564D206, Ectodermal BMP inhibitor, ECTODIN, sclerostin domain containing 1, sclerostin domain-containing protein 1, USAG-1, USAG1uterine sensitization-associated protein-1, Uterine sensitization-associated gene 1 protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human USAG1/SOSTDC1. | |
| 0.1 mL | |
| Signal Transduction | |
| 25928 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction