missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF2 Antibody (PCRP-USF2-1A7) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24186JF635
This item is not returnable.
View return policy
Description
USF2 Monoclonal antibody specifically detects USF2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| USF2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| BHLHB12, bHLHb12upstream stimulatory factor 2, Class B basic helix-loop-helix protein 12, FIPc-fos interacting protein, FOS-interacting protein, Major late transcription factor 2, Upstream transcription factor 2, upstream transcription factor 2, c-fos interacting | |
| Recombinant fragment (aa220-346) of human USF2 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PCRP-USF2-1A7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7392 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction