missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP10 Antibody (OTI2E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74798CL1
This item is not returnable.
View return policy
Description
USP10 Monoclonal antibody specifically detects USP10 in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Gene Knock-Out,Immunocytochemistry/Immunofluorescence
Specifications
| USP10 | |
| Monoclonal | |
| CoraFluor 1 | |
| Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO | |
| Full length human recombinant protein of human USP10(NP_005144) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology | |
| 9100 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Gene Knock-Out | |
| OTI2E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction