missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP10 Antibody (OTI2E1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74798UV
This item is not returnable.
View return policy
Description
USP10 Monoclonal specifically detects USP10 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| USP10 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP10 | |
| Full length recombinant protein of human USP10 (NP_005144) produced in HEK293T cells. | |
| 0.1 mL | |
| 9100.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| OTI2E1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction