missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP11 Antibody (S08-2D7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19545-50ul
This item is not returnable.
View return policy
Description
USP11 Monoclonal antibody specifically detects USP11 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| USP11 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-2D7 | |
| Unconjugated | |
| Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked | |
| A synthetic peptide of human USP11 (Uniprot # P51784) | |
| 50 μg | |
| Cell Biology, Direct Reversal of DNA Damage | |
| 8237 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction