Learn More
USP11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22340-1-AP-150UL
This item is not returnable.
View return policy
Description
USP11 (ubiquitin specific peptidase 11), also known as UHX1, is a 920 amino acid deubiquitinating enzyme that participates in the Ub pathway. Localized to the nucleus, USP11 associates with both Ran BP-M (Ran binding protein M) and with the tumor suppressor BRCA2. Through these associations, USP11 functions to either inhibit ubiquitination of these proteins or to remove ubiquitin residues that have already been attached to these proteins. USP11 is implicated in several X-linked retinal diseases and, due to its ability to deubiquitinate BRCA2, may play a role in tumor suppression.Specifications
| USP11 | |
| Polyclonal | |
| Unconjugated | |
| USP11 | |
| Deubiquitinating enzyme 11, Ubiquitin thiolesterase 11, UHX1, USP11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8237 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51784 | |
| USP11 | |
| USP11 Fusion Protein Ag17829 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |