missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP12 Antibody (OTI1E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46298
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
USP12 Monoclonal specifically detects USP12 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| USP12 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| Deubiquitinating enzyme 12, EC 3.1.2.15, EC 3.4.19.12, UBH1ubiquitin specific protease 12, ubiquitin carboxyl-terminal hydrolase 12, ubiquitin specific peptidase 12, ubiquitin specific protease 12 like 1, ubiquitin thioesterase 12, Ubiquitin thiolesterase 12, Ubiquitin-hydrolyzing enzyme 1, Ubiquitin-specific-processing protease 12, USP12L1 | |
| Mouse | |
| 42.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1E3 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| USP12 | |
| Human recombinant protein fragment corresponding to amino acids 61-251 of human USP12 (NP_872294) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 219333 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction