missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46203PCP
This item is not returnable.
View return policy
Description
USP13 Polyclonal antibody specifically detects USP13 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| USP13 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13 | |
| The immunogen for this product maps to a region between residue 175 and 225 of human Ubiquitin Specific Peptidase 13 using the numbering given in entry NP_003931.2 (GeneID 8975). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8975 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction