Learn More
USP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14354-1-AP-20UL
 View more versions of this product
Product Code. 16892954
  • € 145.00 / 20µL
Description
This gene encodes a member of the ubiquitin specific protease (USP) family of deubiquitinating enzymes. USP enzymes play critical roles in ubiquitin-dependent processes through polyubiquitin chain disassembly and hydrolysis of ubiquitin-substrate bonds. The encoded protein associates with the COP9 signalosome, and also plays a role in transforming growth factor beta signaling through deubiquitination of receptor-activated SMAD transcription factors. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 2.
Specifications
Specifications
USP15
Polyclonal
Unconjugated
USP15
Deubiquitinating enzyme 15, KIAA0529, Ubiquitin thiolesterase 15, Unph 2, UNPH4, USP15
Rabbit
Antigen Affinity Chromatography
RUO
14479, 171329, 9958
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R5H1, Q9R085, Q9Y4E8
Usp15
USP15 Fusion Protein Ag5703
20 μL
Primary
Mouse, Human, Rat