missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ USP16 Antibody [FITC], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-81638F
This item is not returnable.
View return policy
Description
USP16 Polyclonal antibody specifically detects USP16 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| USP16 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| FITC | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 16 using the numbering given in entry NP_006438.1 (GeneID 10600). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10600 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction