missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP16 Antibody (OTI1B4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74805
This item is not returnable.
View return policy
Description
USP16 Monoclonal specifically detects USP16 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.
Specifications
| USP16 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:200-4000 | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 10600 | |
| Human, Mouse, Rat, Canine, Monkey | |
| Lyophilized |
| Western Blot | |
| OTI1B4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| USP16 | |
| Human recombinant protein fragment corresponding to amino acids 137-466 of human USP16(NP_001001992) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction