missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP16 Antibody (OTI1B4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74805UV
This item is not returnable.
View return policy
Description
USP16 Monoclonal specifically detects USP16 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.
Specifications
| USP16 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP16 | |
| Recombinant protein fragment corresponding to amino acids 137-466 of human USP16 (NP_001001992) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot | |
| OTI1B4 | |
| Western Blot | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 10600.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction