missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP16 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596452

Product Code. 16539291

  • € 528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MGKKRTKGKT VPIDDSSETL EPVCRHIRKG LEQGNLKKAL VNVEWNICQD CKTDNKVKDK AEEETEEKPS VWLCLKCGHQ GCGRNSQEQH ALKHYLTPRS EPHCLVLSLD NWSVWCYVCD NEVQYCSSNQ LGQVVDYVRK QASITTPKPE KDNGNIELEN KKLEKESKNE QEREKKENMA KENPPMNSPC QITVKGLSNL GNTCFFNAVM QNLSQTPVLR ELLKEVKMSG TIVKIEPPDL ALTEPLEINL EPPGPLTLAM SQFLNEMQET KKGVVTPKEL; Positive Samples: COS-7, 293T, NIH/3T3, Mouse brain, Mouse heart, Mouse spleen, Rat brain; Cellular Location: Nucleus.

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP16
Polyclonal
Unconjugated
USP16
1200004E02Rik; 2810483I07Rik; 6330514E22Rik; Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255; HAMAP-Rule:MF_03062}; MSTP039; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255; Ubiquitin carboxyl-terminal hydrolase 16-like protein; ubiquitin specific peptidase 16; ubiquitin specific protease 16; ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255; ubiquitin thiolesterase 16; ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255; UBPM; UBP-M; USP16
Rabbit
Affinity chromatography
RUO
10600, 288306, 74112
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.67 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q2KJ09, Q99LG0, Q9Y5T5
USP16
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human USP16 (NP_0010019921).
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.