missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP17L3 Polyclonal Antibody

Product Code. 15587232
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15587232 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15587232 Supplier Invitrogen™ Supplier No. PA546659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: SESVSRGREP RALGAEDTDR RAKQGELKRD HPCLQAPELD EHLVERATQE Sequence homology: Human: 100%.

USP17L3 is a deubiquitinating enzyme that removes conjugated ubiquitin from specific proteins to regulate different cellular processes that may include cell proliferation, progression through the cell cycle, apoptosis, cell migration, and the cellular response to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP17L3
Gene Accession No. A6NCW0
Gene Alias Deubiquitinating enzyme 17-like protein 3; ubiquitin carboxyl-terminal hydrolase 17-like protein 3; ubiquitin specific peptidase 17-like 3; ubiquitin specific peptidase 17-like family member 3; ubiquitin thioesterase 17-like protein 3; Ubiquitin-specific-processing protease 17-like protein 3; USP17B; USP17F; USP17L3
Gene Symbols USP17L3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human USP17L3 (aa 382-431).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645836
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.