missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | USP18 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230596
|
Novus Biologicals
NBP3-35697-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228983
|
Novus Biologicals
NBP3-35697-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP18 Polyclonal antibody specifically detects USP18 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| USP18 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 11274 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1.2.15, EC 3.4.19.-, hUBP43, ISG15-specific-processing protease, ISG43UBP43, ubiquitin specific peptidase 18, ubiquitin specific protease 18, ubl carboxyl-terminal hydrolase 18, ubl thioesterase 18,43 kDa ISG15-specific protease, Ubl thiolesterase 18 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human USP18 (NP_059110.2).,, Sequence:, DVDSKPLKTLEDALHCFFQPRELSSKSKCFCENCGKKTRGKQVLKLTHLPQTLTIHLMRFSIRNSQTRKICHSLYFPQSLDFSQILPMKRESCDAEEQSGG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title