missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598234
This item is not returnable.
View return policy
Description
USP2 Polyclonal Antibody for ICC/IF, ELISA
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
Specifications
| USP2 | |
| Polyclonal | |
| Unconjugated | |
| USP2 | |
| 41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9099 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.273 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O75604 | |
| USP2 | |
| Recombinant Human Ubiquitin carboxyl-terminal hydrolase 2 protein (9-265AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction