missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13454769
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454769 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454769 Supplier Invitrogen™ Supplier No. PA533099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bovine, Hamster, Elephant, Panda, Horse, Rabbit, Opossum (100%); Pig (94%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.

Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene USP2
Gene Accession No. O75604, O88623, Q2KHV7, Q5U349
Gene Alias 41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9
Gene Symbols USP2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-terminus of human USP2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100071440, 100339033, 100758523, 115771, 522980, 53376, 608188, 9099
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.