missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15404-1-AP-150UL
This item is not returnable.
View return policy
Description
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).Specifications
| USP2 | |
| Polyclonal | |
| Unconjugated | |
| USP2 | |
| Deubiquitinating enzyme 2, ubiquitin specific peptidase 2, Ubiquitin thiolesterase 2, UBP41, USP2, USP9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115771, 53376, 9099 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75604, O88623, Q5U349 | |
| Usp2 | |
| USP2 Fusion Protein Ag7658 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction