missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17856-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described.Specifications
| USP21 | |
| Polyclonal | |
| Unconjugated | |
| USP21 | |
| Deubiquitinating enzyme 21, Ubiquitin thiolesterase 21, USP16, USP21, USP23 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27005 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UK80 | |
| USP21 | |
| USP21 Fusion Protein Ag12316 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction