Learn More
USP22 Antibody (18E3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26260-100ul
 View more versions of this product
Product Code. 30029716
  • € 533.00 / 100µL
Description
USP22 Monoclonal antibody specifically detects USP22 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP22
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, Immunocytochemistry
18E3
Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L
A synthesized peptide derived from Human USP22 [UniProt Q9UPT9]
100 μL
Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers
23326
Store at -20 to -70C. Avoid freeze-thaw cycles.