missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP25 Monoclonal Antibody (1277CT376.106.171)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537640
This item is not returnable.
View return policy
Description
USP25 Monoclonal Antibody for Western Blot
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
Specifications
| USP25 | |
| Monoclonal | |
| 0.49 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UHP3 | |
| Usp25 | |
| recombinant protein from human USP25. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 1277CT376.106.171 | |
| Unconjugated | |
| Usp25 | |
| Deubiquitinating enzyme 25; mUSP25; Ubiquitin carboxyl-terminal hydrolase 25; ubiquitin specific peptidase 25; ubiquitin specific protease 25; ubiquitin thioesterase 25; ubiquitin thiolesterase 25; ubiquitin-specific processing protease 25; ubiquitin-specific-processing protease 25; USP on chromosome 21; USP21; Usp25 | |
| Mouse | |
| Protein G | |
| RUO | |
| 29761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction