missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104441
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP29.
USP29 (Ubiquitin Specific Peptidase 29) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitin-specific protease activity. An important paralog of this gene is USP37. [GeneCards].
Specifications
| USP29 | |
| Polyclonal | |
| Unconjugated | |
| USP29 | |
| ASL1/Usp29 fusion; deubiquitinating enzyme 29; HOM-TES-84/86; Ocat; ossification center-associated transcript; ubiquitin carboxyl-terminal hydrolase 29; ubiquitin specific peptidase 29; ubiquitin specific protease 29; ubiquitin thioesterase 29; ubiquitin thiolesterase 29; ubiquitin-specific processing protease; ubiquitin-specific-processing protease 29; Usp29 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361495, 57663, 57775 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ES63, Q9HBJ7 | |
| USP29 | |
| A synthesized peptide derived from human USP29(Accession Q9HBJ7), corresponding to amino acid residues Q692-I742. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction