missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP3 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77284R
This item is not returnable.
View return policy
Description
USP3 Polyclonal antibody specifically detects USP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| USP3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Deubiquitinating enzyme 3, EC 3.1.2.15, EC 3.4.19.12, MGC129878, MGC129879, SIH003, ubiquitin carboxyl-terminal hydrolase 3, ubiquitin specific peptidase 3, ubiquitin specific protease 3, ubiquitin thioesterase 3, Ubiquitin thiolesterase 3, Ubiquitin-specific-processing protease 3, UBP | |
| The immunogen recognized by this antibody maps to a region between residue 471 and 521 of human ubiquitin specific protease 3 using the numbering given in entry AAD42992.1 (GeneID 9960). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9960 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction