missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111372
This item is not returnable.
View return policy
Description
Immunogen sequence: CCYFKELPAV ELRNGKTAGR RTYHTRSQGD NNVSLVEEFR KTLCALWQGS QTAFSPESLF YVVWKIMPNF RGYQ Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
Specifications
| USP3 | |
| Polyclonal | |
| Unconjugated | |
| USP3 | |
| AA409661; BC017156; Deubiquitinating enzyme 3; SIH003; Ubiquitin carboxyl-terminal hydrolase 3; ubiquitin specific peptidase 3; ubiquitin specific protease 3; ubiquitin thioesterase 3; ubiquitin thiolesterase 3; ubiquitin-specific-processing protease 3; UBP; Usp3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6I4 | |
| USP3 | |
| Recombinant Protein corresonding to Human USP3. Recombinant protein control fragment (Product #RP-106448). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction