missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP30 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17140914 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140914 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17140914 Supplier Invitrogen™ Supplier No. PA5113053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP30 Polyclonal Antibody for IHC (P), ELISA

USP30, a member of the ubiquitin-specific protease family, is a novel mitochondrial deubiquitinating (DUB) enzyme (Nakamura and Hirose, 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP30
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene USP30
Gene Accession No. Q70CQ3
Gene Alias 6330590F17Rik; AI851327; D5Ertd483e; Deubiquitinating enzyme 30; FLJ40511; MGC10702; Ubiquitin carboxyl-terminal hydrolase 30; ubiquitin specific peptidase 30; ubiquitin specific protease 30; ubiquitin thioesterase 30; ubiquitin thiolesterase 30; ubiquitin-specific protease 30; ubiquitin-specific-processing protease 30; ub-specific protease 30; USP30
Gene Symbols USP30
Host Species Rabbit
Immunogen Recombinant Human Ubiquitin carboxyl-terminal hydrolase 30 protein (101-400AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84749
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.