missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106447
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total USP33.
DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
Specifications
| USP33 | |
| Polyclonal | |
| Unconjugated | |
| USP33 | |
| 9830169D19Rik; AA409780; Deubiquitinating enzyme 33; hVDU1; KIAA1097; pVHL-interacting deubiquitinating enzyme 1; ubiquitin carboxyl-terminal hydrolase 33; ubiquitin specific peptidase 33; ubiquitin specific protease 33; ubiquitin thioesterase 33; ubiquitin thiolesterase 33; ubiquitin-specific-processing protease 33; Usp33; VDU1; Vhlh-interacting deubiquitinating enzyme 1; VHL-interacting deubiquitinating enzyme 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170822, 23032, 310960 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R5K2, Q8TEY7 | |
| USP33 | |
| A synthesized peptide derived from human USP33(Accession Q8TEY7), corresponding to amino acid residues M292-D342. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction