missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP36 Antibody (OTI7G3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74806FR
This item is not returnable.
View return policy
Description
USP36 Monoclonal specifically detects USP36 in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| USP36 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| USP36 | |
| Recombinant protein fragment corresponding to amino acids 589-972 of human USP36 (NP_079366) produced in E.coli. | |
| 0.1 mL | |
| 57602.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI7G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Deubiquitinating enzyme 36, DUB1, EC 3.1.2.15, EC 3.4.19.12, FLJ12851, KIAA1453deubiquitinating enzyme 1, ubiquitin carboxyl-terminal hydrolase 36, ubiquitin specific peptidase 36, ubiquitin specific protease 36, ubiquitin thioesterase 36, Ubiquitin thiolesterase 36, Ubiquitin-specific-processing protease 36 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction