missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP37 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40696CL1
This item is not returnable.
View return policy
Description
USP37 Polyclonal antibody specifically detects USP37 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot (Negative)
Specifications
| USP37 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37 | |
| The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ubiquitin Specific Peptidase 37 using the numbering given in entry NP_065986.1 (GeneID 57695). | |
| 0.1 mL | |
| Protein Turnover, Ubiquitin Proteasome Pathway | |
| 57695 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction