missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142059
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| USP4 | |
| Polyclonal | |
| Unconjugated | |
| USP4 | |
| Deubiquitinating enzyme 4; F730026I20Rik; mKIAA4155; ubiquitin carboxyl-terminal esterase 4; ubiquitin carboxyl-terminal hydrolase 4; ubiquitin specific peptidase 4; ubiquitin specific peptidase 4 (proto-oncogene); ubiquitin specific protease 4 (proto-oncogene); ubiquitin thioesterase 4; ubiquitin thiolesterase 4; ubiquitin-specific processing protease 4; ubiquitin-specific-processing protease 4; Ubiquitous nuclear protein; ubiquitous nuclear protein homolog; Unp; Unph; USP4 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7375 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13107 | |
| USP4 | |
| Peptide with sequence AEGGGCRERPDAE (aa 2-14). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction