missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP44 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801912
This item is not returnable.
View return policy
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.Specifications
| USP44 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H0E7 | |
| USP44 | |
| Human recombit protein fragment corresponding to amino acids 420-712 of human USP44 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI1C2 | |
| Unconjugated | |
| USP44 | |
| ubiquitin specific peptidase 44 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction