missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP44 Monoclonal Antibody (OTI5F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801908S
This item is not returnable.
View return policy
Description
USP44 Monoclonal antibody specifically detects USP44 in Human samples. It is validated for Western BlotSpecifications
| USP44 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62760 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7447 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5F2 | |
| Unconjugated | |
| VSNL1 | |
| Deubiquitinating enzyme 44; E430004F17Rik; hypothetical protein MGC55661; inactive deubiquitinating enzyme 44; inactive ubiquitin carboxyl-terminal hydrolase 44; inactive ubiquitin thioesterase 44; inactive ubiquitin-specific-processing protease 44; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 44; ubiquitin carboxyl-terminal hydrolase 44; ubiquitin specific peptidase 44; ubiquitin specific peptidase 49 pseudogene; ubiquitin specific protease 44; ubiquitin thioesterase 44; ubiquitin thiolesterase 44; ubiquitin-specific-processing protease 44; USP44; zgc:55661 | |
| Human recombinant protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction