Learn More
USP44 Mouse anti-Human, Clone: OTI1A9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF801917
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.Specifications
| USP44 | |
| Monoclonal | |
| Unconjugated | |
| USP44 | |
| ubiquitin specific peptidase 44 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H0E7 | |
| USP44 | |
| Human recombit protein fragment corresponding to amino acids 420-712 of human USP44 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.