missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99882-50ul
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
USP46 Polyclonal antibody specifically detects USP46 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Tekniske data
| USP46 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Deubiquitinating enzyme 46, EC 3.1.2.15, EC 3.4.19.12, FLJ11850, FLJ12552, FLJ14283, FLJ39393, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific peptidase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, Ubiquitin thiolesterase 46, Ubiquitin-specific-processing protease 46 | |
| Produced in rabbits immunized with purified, recombinant Human/Mouse USP46 (Accession#: NP_808229.1; Met1-Glu366). | |
| 50 μg | |
| DNA replication Transcription Translation and Splicing | |
| 64854 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion