missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP47 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13217818
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13217818 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13217818 Supplier Invitrogen™ Supplier No. PA521732

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (89%), Rat (92%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ubiquitin-specific protease that specifically deubiquitinates monoubiquitinated DNA polymerase beta (POLB), stabilizing POLB thereby playing a role in base-excision repair (BER). Acts as a regulator of cell growth and genome integrity. May also indirectly regulate CDC25A expression at a transcriptional level. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP47
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene USP47
Gene Accession No. Q8BY87, Q96K76
Gene Alias 4930502N04Rik; A630020C16Rik; deubiquitinating enzyme 47; Trf (TATA binding protein-related factor)-proximal homolog; TRFP; Ubiquitin carboxyl-terminal hydrolase 47; ubiquitin specific peptidase 47; ubiquitin specific protease 47; ubiquitin thioesterase 47; ubiquitin thiolesterase 47; ubiquitin-specific-processing protease 47; Usp47
Gene Symbols USP47
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP47. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55031, 74996
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.