missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP48 Antibody (OTI1F10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44676
This item is not returnable.
View return policy
Description
USP48 Monoclonal antibody specifically detects USP48 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| USP48 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 84196 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| OTI1F10 | |
| Western Blot 1:500, Immunohistochemistry 1:150 | |
| Deubiquitinating enzyme 48, DKFZp762M1713, EC 3.1.2.15, EC 3.4.19.12, FLJ11328, FLJ20103, FLJ23054, FLJ23277, MGC132556, MGC14879, RAP1GA1, ubiquitin carboxyl-terminal hydrolase 48, ubiquitin specific peptidase 48, ubiquitin specific protease 31, ubiquitin specific protease 48, ubiquitin thioesterase 48, Ubiquitin thiolesterase 48, Ubiquitin-specific-processing protease 48, USP31 | |
| Human recombinant protein fragment corresponding to amino acids 361-743 of human USP48 (NP_115612) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction