missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP5 Monoclonal Antibody (OTI1F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501287
This item is not returnable.
View return policy
Description
USP5 (ubiquitin carboxyl-terminal hydrolase 5) cleaves linear and branched multiubiquitin polymers with a marked preference for branched polymers. It is involved in uchored Lys-48-linked polyubiquitin disassembly, and also binds linear and Lys-63 linked polyubiquitin with a lower affinity. Knock-down of USP5 causes the accumulation of p53/TP53 and an increase in p53/TP53 transcriptional activity because the uchored polyubiquitin that accumulates is able to compete with ubiquitinated p53/TP53 but not with MDM2 for proteasomal recognition.Specifications
| USP5 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P45974 | |
| USP5 | |
| Full length human recombit protein of human USP5 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F8 | |
| Unconjugated | |
| USP5 | |
| ISOT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 297593, 486718, 8078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction