missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP50 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15746671 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15746671 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15746671 Supplier Invitrogen™ Supplier No. PA558799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YHYSRRRSYE KGSTQRCCRK WITTETSIIT QLFEEQLNYS IVCLKCEKCT YKNEVFTVFS LPIPSKYECS LRDCLQCF Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

Has no peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP50
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Usp50
Gene Accession No. Q70EL3
Gene Alias 1700086G18Rik; 4930511O11Rik; Deubiquitinating enzyme 50; inactive ubiquitin carboxyl-terminal hydrolase 50; Inactive ubiquitin-specific peptidase 50; putative ubiquitin carboxyl-terminal hydrolase 50; RGD1564751; ubiquitin specific peptidase 50; ubiquitin specific protease 50; ubiquitin thioesterase 50; ubiquitin thiolesterase 50; ubiquitin-specific-processing protease 50; USP50
Gene Symbols Usp50
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP50. Recombinant protein control fragment (Product #RP-95857).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.