missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP53 Antibody (OTI3C9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74812G
This item is not returnable.
View return policy
Description
USP53 Monoclonal specifically detects USP53 in Human samples. It is validated for Western Blot.
Specifications
| USP53 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP53 | |
| Recombinant protein fragment corresponding to amino acids 495-582 of human USP53 (NP_061923) produced in E.coli. | |
| 0.1 mL | |
| 54532.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3C9 | |
| Western Blot | |
| DKFZp781E1417, inactive ubiquitin carboxyl-terminal hydrolase 53, Inactive ubiquitin-specific peptidase 53, KIAA1350, ubiquitin specific peptidase 53, ubiquitin specific protease 53, ubiquitin specific proteinase 53 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction