missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP54 Antibody (OTI1G6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74813R
This item is not returnable.
View return policy
Description
USP54 Monoclonal specifically detects USP54 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| USP54 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| USP54 | |
| Recombinant protein fragment corresponding to amino acids 358-700 of human USP54 (NP_689799) produced in E.coli. | |
| 0.1 mL | |
| 159195.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1G6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| bA137L10.3, bA137L10.4, C10orf29, USP54 ubiquitin specific peptidase 54 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction