missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP54 Antibody (OTI1G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45403
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunofluorescence
USP54 Monoclonal specifically detects USP54 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| USP54 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| bA137L10.3, bA137L10.4, C10orf29, USP54 ubiquitin specific peptidase 54 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 159195 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1G6 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| USP54 | |
| Human recombinant protein fragment corresponding to amino acids 358-700 of human USP54 (NP_689799) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction