missing translation for 'onlineSavingsMsg'
Learn More

USP7/USP47 inhibitor, MedChemExpress

Product Code. 16476196
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476196 100 mg 100mg
16488397 5 mg 5mg
16471867 50 mg 50mg
16455708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476196 Supplier Medchem Express Supplier No. HY13487

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress USP7/USP47 inhibitor is a selective ubiquitin-specific protease 7/47 (USP7/USP47) inhibitor, with EC50s of 0.42 μM and 1.0 μM, respectively.

USP7/USP47 inhibitor is a selective ubiquitin-specific protease 7/47 (USP7/USP47) inhibitor, with EC50s of 0.42 μM and 1.0 μM, respectively.

Specifications

Chemical Name or Material USP7/USP47 inhibitor
CAS 1247825-37-1
Color White
Purity Grade Notes Research
Molecular Formula C18H11Cl2N3O3S3
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (103.22 mM; Need ultrasonic)
SMILES ClC1=CN=CC(Cl)=C1SC2=C(C#N)C=C(C(NC3=CC=C(S(C)(=O)=O)C=C3)=O)S2
Molecular Weight (g/mol) 484.4
Formula Weight 484.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.