missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP9X, Mouse anti-Human, Clone: 2A9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008239-M08.50ug
This item is not returnable.
View return policy
Description
Sequence: MTATTRGSPVGGNDNQGQAPDGQSQPPLQQNQTSSPDSSNENSPATPPDEQGQGDAPPQLEDEEPAFPHTDLAKLDDMINRPRWVVPVLPSpecifications
| USP9X | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin specific peptidase 9, X-linked | |
| DFFRX/FAF/FAM | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A9 | |
| In 1x PBS, pH 7.4 | |
| NM_021906 | |
| USP9X | |
| USP9X (NP_068706, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8239 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction