Learn More
Invitrogen™ USP9Y Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570428
Product Code. 15888833
  • € 526.00 / 100µL
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%.
USP9Y gene ontology annotations related to this gene include protein deubiquitination; ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP9Y
Polyclonal
Unconjugated
USP9Y
Deubiquitinating enzyme FAF-Y; DFFRY; Fafl2; Fat facets protein-related, Y-linked; probable ubiquitin carboxyl-terminal hydrolase FAF-X; probable ubiquitin carboxyl-terminal hydrolase FAF-Y; SPGFY2; ubiquitin specific peptidase 9, Y chromosome; ubiquitin specific peptidase 9, Y-linked; ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila); ubiquitin specific protease 9, Y chromosome; ubiquitin thioesterase FAF-Y; ubiquitin thiolesterase FAF-Y; ubiquitin-specific processing protease FAF-Y; ubiquitin-specific protease 9, Y chromosome; Ubiquitin-specific-processing protease FAF-Y; USP9Y
Rabbit
Affinity chromatography
RUO
8287
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O00507
USP9Y
synthetic peptide directed towards the C terminal region of human USP9Y.