missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USPL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15726621 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15726621 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15726621 Supplier Invitrogen™ Supplier No. PA558745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YCPACREKGK LKALKTYRIS FQESIFLCED LQCIYPLGSK SLNNLISPDL EECHTPHKPQ KRKSLESSYK DSLLLANSKK TRNYIAIDGG KVLNSKHNGE VYDETSSNLP DSSGQQNP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen USPL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USPL1
Gene Accession No. Q5W0Q7
Gene Alias bA121O19.1; C13orf22; D13S106; D13S106E; highly charged protein; SUMO-specific isopeptidase USPL1; ubiquitin specific peptidase like 1; ubiquitin-specific peptidase-like protein 1; USPL1; USP-like 1
Gene Symbols USPL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USPL1. Recombinant protein control fragment (Product #RP-97951).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.