missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (016), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90515AF405
This item is not returnable.
View return policy
Description
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Mouse samples. It is validated for ELISA.
Specifications
| Uteroglobin/SCGB1A1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Uteroglobin/SCGB1A1 (Q06318; Met1-Phe96). | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 7356 | |
| Mouse | |
| Purified |
| ELISA | |
| 16 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Human SCGB1A1 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction