missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTF1 Antibody (5B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 217.00 - € 557.00
Specifications
| Antigen | UTF1 |
|---|---|
| Clone | 5B6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625215
|
Novus Biologicals
NBP2-37444-0.025ml |
0.025 mL |
€ 217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18161199
|
Novus Biologicals
NBP2-37444 |
0.1 mL |
€ 557.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UTF1 Monoclonal specifically detects UTF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| UTF1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| undifferentiated embryonic cell transcription factor 1 | |
| UTF1 | |
| IgG1 | |
| Affinity Purified |
| 5B6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 8433 | |
| Purified recombinant fragment of human UTF1 (AA: 148-214) expressed in E. Coli. | |
| Primary | |
| 36.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title