missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ uTIP™ Sterile Low Retention Universal Pipette Tips, Standard
GREENER_CHOICE

Product Code. 30216531
Click to view available options
Volume (Metric):
250 μL
1000 μL
Unit Size:
Pack of 960

Product Code. 30216531

Brand: BIOTIX™ 63300051

Please to purchase this item. Need a web account? Register with us today!

Edge

Designed with FlexFit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Non-Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.

  • Compatible with the widest range of industry leading pipettes - Fits all standard pipettes
  • Accurate and precise channel-channel draw on multichannel pipettes
  • Secure seal on single and multichannel models
  • FlexFit™ seals on your pipette with less force, reducing your ejection force
  • FlexFit™ technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin™ maximizes your sample recovery by creating a low binding surface for DNA, proteins, and nanoparticles - resulting in tighter CV values
  • Blade™ lowers chance of hanging droplet formation by optimizing surface area at distal end of tip - ensures a clean delivery of your sample, increasing accuracy and precision
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
  • Better reprodicibility
  • Certifications

    ISO 9001

    Specifications

    Description 250 μL, Racked, Sterile
    Volume (Metric) 250 μL
    Filtered Non-filtered
    Sterility Sterile
    Color Clear
    Rack Color Green
    Autoclavable Autoclavable
    Model Universal
    Length (Metric) 59.54 mm
    Certifications/Compliance ISO 9001
    Clean Claims Reusable as a 96 well reservoir
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Sterilization Method e-beam
    Format Hinged Rack
    Tip Style Ergonomic
    Packaging Racked
    Quantity 10 racks/pk
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line uTip
    Material Polypropylene
    Shape Conical
    Graduations 10 μL, 50 μL, 100 μL, 200 μL
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.