missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP11L Antibody (OTI5F3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74823B
This item is not returnable.
View return policy
Description
UTP11L Monoclonal specifically detects UTP11L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UTP11L | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| UTP11L | |
| Full length recombinant protein of human UTP11L (NP_057121) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5F3 | |
| Western Blot, Immunohistochemistry | |
| CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Cytokine Research | |
| 51118.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction