missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP11L Antibody (OTI5F3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74823CL1
This item is not returnable.
View return policy
Description
UTP11L Monoclonal antibody specifically detects UTP11L in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| UTP11L | |
| Monoclonal | |
| CoraFluor 1 | |
| CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
| Full length human recombinant protein of human UTP11L(NP_057121) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, Cytokine Research | |
| 51118 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI5F3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction